Assay Details
Gene Symbol: LOC5571585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849062.1 | XP_021704754.1 | 3 - 4 | 1748 | 58 | |
XM_021849063.1 | XP_021704755.1 | 3 - 4 | 1787 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
