Assay Details
Gene Symbol: LOC5567090 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841756.1 | XP_021697448.1 | 6 - 7 | 2988 | 82 | |
XM_021841757.1 | XP_021697449.1 | 6 - 7 | 2920 | 82 | ||
XM_021841758.1 | XP_021697450.1 | 6 - 7 | 2973 | 82 | ||
XM_021841759.1 | XP_021697451.1 | 6 - 7 | 2905 | 82 |
