Assay Details
Gene Symbol: LOC5567090 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841756.1 | XP_021697448.1 | 3 - 4 | 550 | 68 | |
XM_021841757.1 | XP_021697449.1 | 3 - 4 | 482 | 68 |

More Information
Set Membership: |
![]() ![]() |
