Assay Details
Gene Symbol: LOC5567090 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841756.1 | XP_021697448.1 | 2 - 3 | 368 | 62 | |
XM_021841757.1 | XP_021697449.1 | 2 - 3 | 300 | 62 | ||
XM_021841758.1 | XP_021697450.1 | 2 - 3 | 368 | 62 | ||
XM_021841759.1 | XP_021697451.1 | 2 - 3 | 300 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
