Assay Details
Gene Symbol: LOC5564923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848061.1 | XP_021703753.1 | 1 - 2 | 143 | 114 | |
XM_021848065.1 | XP_021703757.1 | 1 - 2 | 148 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021848061.1, XM_021848065.1 transcripts.
Set Membership: |
![]() ![]() |
