Assay Details
Gene Symbol: LOC5564923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848061.1 | XP_021703753.1 | 9 - 10 | 1673 | 83 | |
XM_021848062.1 | XP_021703754.1 | 9 - 10 | 1703 | 83 | ||
XM_021848063.1 | XP_021703755.1 | 9 - 10 | 1650 | 83 | ||
XM_021848065.1 | XP_021703757.1 | 9 - 10 | 1678 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
