Assay Details
Gene Symbol: LOC5573640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660916.3 | XP_001660966.3 | 2 - 3 | 1311 | 62 | |
XM_021837700.1 | XP_021693392.1 | 2 - 3 | 1311 | 62 | ||
GenBank mRNA | KC439539.1 | - | 1 - 2 | 254 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
