Assay Details
Gene Symbol: LOC5572154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660178.2 | XP_001660228.2 | 2 - 3 | 469 | 104 | |
XM_001660179.2 | XP_001660229.2 | 2 - 3 | 476 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001660178.2 transcripts.
Set Membership: |
![]() ![]() |
