Assay Details
Gene Symbol: LOC5568995 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849078.1 | XP_021704770.1 | 13 - 14 | 3018 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5568995 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849078.1 | XP_021704770.1 | 13 - 14 | 3018 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() |