Assay Details
Gene Symbol: LOC5570958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659376.2 | XP_001659426.1 | 1 - 2 | 190 | 66 | |
XM_021845805.1 | XP_021701497.1 | 1 - 2 | 190 | 66 | ||
XM_021845806.1 | XP_021701498.1 | 1 - 2 | 174 | 66 | ||
GenBank mRNA | KJ598054.1 | - | 1 - 2 | 195 | 66 | |
KJ598055.1 | - | 1 - 2 | 195 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001659376.2, XM_021845805.1, XM_021845806.1 transcripts.
Set Membership: |
![]() ![]() |
