Assay Details
Gene Symbol: LOC5577549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847099.1 | XP_021702791.1 | 2 - 3 | 422 | 91 | |
XM_021847100.1 | XP_021702792.1 | 2 - 3 | 365 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021847099.1, XM_021847100.1 transcripts.
Set Membership: |
![]() ![]() |
