Assay Details
Gene Symbol: LOC5577549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847098.1 | XP_021702790.1 | 2 - 3 | 1013 | 98 | |
XM_021847099.1 | XP_021702791.1 | 3 - 4 | 1119 | 98 | ||
XM_021847100.1 | XP_021702792.1 | 3 - 4 | 1062 | 98 | ||
XM_021847101.1 | XP_021702793.1 | 2 - 3 | 918 | 98 | ||
XM_021847103.1 | XP_021702795.1 | 2 - 3 | 836 | 98 |
