Assay Details
Gene Symbol: LOC5577549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847098.1 | XP_021702790.1 | 6 - 7 | 2682 | 92 | |
XM_021847099.1 | XP_021702791.1 | 7 - 8 | 2788 | 92 | ||
XM_021847100.1 | XP_021702792.1 | 7 - 8 | 2731 | 92 | ||
XM_021847101.1 | XP_021702793.1 | 6 - 7 | 2587 | 92 | ||
XM_021847103.1 | XP_021702795.1 | 6 - 7 | 2505 | 92 |
