Assay Details
Gene Symbol: LOC5577549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847098.1 | XP_021702790.1 | 7 - 8 | 2897 | 93 | |
XM_021847099.1 | XP_021702791.1 | 8 - 9 | 3003 | 93 | ||
XM_021847100.1 | XP_021702792.1 | 8 - 9 | 2946 | 93 | ||
XM_021847101.1 | XP_021702793.1 | 7 - 8 | 2802 | 93 | ||
XM_021847103.1 | XP_021702795.1 | 7 - 8 | 2720 | 93 |
