Assay Details
Gene Symbol: LOC5577549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847098.1 | XP_021702790.1 | 8 - 9 | 3141 | 75 | |
XM_021847099.1 | XP_021702791.1 | 9 - 10 | 3247 | 75 | ||
XM_021847100.1 | XP_021702792.1 | 9 - 10 | 3190 | 75 | ||
XM_021847101.1 | XP_021702793.1 | 8 - 9 | 3046 | 75 | ||
XM_021847103.1 | XP_021702795.1 | 8 - 9 | 2964 | 75 |
