Assay Details
Gene Symbol: LOC5564179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648449.2 | XP_001648499.1 | 2 - 3 | 826 | 65 | |
XM_021853567.1 | XP_021709259.1 | 3 - 4 | 624 | 65 | ||
XM_021853568.1 | XP_021709260.1 | 3 - 4 | 594 | 65 | ||
XM_021853569.1 | XP_021709261.1 | 3 - 4 | 348 | 65 | ||
XM_021853570.1 | XP_021709262.1 | 2 - 3 | 830 | 65 |

More Information
Set Membership: |
![]() ![]() |
