Assay Details
Gene Symbol: LOC5564179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648449.2 | XP_001648499.1 | 3 - 4 | 931 | 84 | |
XM_021853567.1 | XP_021709259.1 | 4 - 5 | 729 | 84 | ||
XM_021853568.1 | XP_021709260.1 | 4 - 5 | 699 | 84 | ||
XM_021853569.1 | XP_021709261.1 | 4 - 5 | 453 | 84 | ||
XM_021853570.1 | XP_021709262.1 | 3 - 4 | 935 | 84 |
