Assay Details
Gene Symbol: LOC5564179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648449.2 | XP_001648499.1 | 5 - 6 | 1159 | 103 | |
XM_021853567.1 | XP_021709259.1 | 6 - 7 | 957 | 103 | ||
XM_021853568.1 | XP_021709260.1 | 6 - 7 | 927 | 103 | ||
XM_021853569.1 | XP_021709261.1 | 6 - 7 | 681 | 103 |

More Information
Set Membership: |
![]() ![]() |
