Assay Details
Gene Symbol: LOC5564179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648449.2 | XP_001648499.1 | 7 - 8 | 1451 | 61 | |
XM_021853567.1 | XP_021709259.1 | 8 - 9 | 1249 | 61 | ||
XM_021853568.1 | XP_021709260.1 | 8 - 9 | 1219 | 61 | ||
XM_021853569.1 | XP_021709261.1 | 8 - 9 | 973 | 61 | ||
XM_021853570.1 | XP_021709262.1 | 7 - 8 | 1455 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
