Assay Details
Gene Symbol: LOC5579114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657263.2 | XP_001657313.2 | 4 - 5 | 1176 | 74 | |
XM_021855580.1 | XP_021711272.1 | 4 - 5 | 1157 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
