Assay Details
Gene Symbol: LOC5574468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655078.3 | XP_001655128.2 | 2 - 3 | 1238 | 70 | |
XM_021843922.1 | XP_021699614.1 | 2 - 3 | 1244 | 70 | ||
XM_021843923.1 | XP_021699615.1 | 3 - 4 | 588 | 70 | ||
XM_021843924.1 | XP_021699616.1 | 3 - 4 | 525 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
