Assay Details
Gene Symbol: LOC5574468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655078.3 | XP_001655128.2 | 1 - 2 | 1114 | 93 | |
XM_021843922.1 | XP_021699614.1 | 1 - 2 | 1120 | 93 | ||
XM_021843923.1 | XP_021699615.1 | 2 - 3 | 464 | 93 | ||
XM_021843924.1 | XP_021699616.1 | 2 - 3 | 401 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
