Assay Details
Gene Symbol: LOC5569240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857577.1 | XP_021713269.1 | 9 - 10 | 3273 | 66 | |
XM_021857578.1 | XP_021713270.1 | 9 - 10 | 3234 | 66 | ||
XM_021857580.1 | XP_021713272.1 | 9 - 10 | 2925 | 66 | ||
XM_021857581.1 | XP_021713273.1 | 9 - 10 | 2754 | 66 | ||
XM_021857582.1 | XP_021713274.1 | 9 - 10 | 2829 | 66 |

More Information
Set Membership: |
![]() ![]() |
