Assay Details
Gene Symbol: LOC5573299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843374.1 | XP_021699066.1 | 6 - 7 | 3094 | 97 | |
XM_021843375.1 | XP_021699067.1 | 6 - 7 | 3190 | 97 | ||
XM_021843376.1 | XP_021699068.1 | 5 - 6 | 2532 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
