Assay Details
Gene Symbol: LOC5573299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843374.1 | XP_021699066.1 | 3 - 4 | 1844 | 109 | |
XM_021843375.1 | XP_021699067.1 | 3 - 4 | 1940 | 109 | ||
XM_021843376.1 | XP_021699068.1 | 2 - 3 | 1282 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
