Assay Details
Gene Symbol: LOC5573853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845596.1 | XP_021701288.1 | 2 - 3 | 770 | 82 | |
XM_021845597.1 | XP_021701289.1 | 2 - 3 | 841 | 82 | ||
XM_021845598.1 | XP_021701290.1 | 2 - 3 | 623 | 82 | ||
XM_021845599.1 | XP_021701291.1 | 2 - 3 | 586 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
