Assay Details
Gene Symbol: LOC5574326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661307.2 | XP_001661357.2 | 2 - 3 | 566 | 75 | |
XM_021838898.1 | XP_021694590.1 | 2 - 3 | 865 | 75 | ||
XM_021838899.1 | XP_021694591.1 | 2 - 3 | 1140 | 75 | ||
XM_021838900.1 | XP_021694592.1 | 2 - 3 | 1040 | 75 | ||
XM_021838901.1 | XP_021694593.1 | 2 - 3 | 835 | 75 | ||
XM_021838902.1 | XP_021694594.1 | 2 - 3 | 741 | 75 | ||
XM_021838903.1 | XP_021694595.1 | 2 - 3 | 620 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001661307.2, XM_021838898.1, XM_021838899.1, XM_021838900.1, XM_021838901.1, XM_021838902.1, XM_021838903.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
