Assay Details
Gene Symbol: LOC5574326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661307.2 | XP_001661357.2 | 4 - 5 | 942 | 63 | |
XM_021838898.1 | XP_021694590.1 | 4 - 5 | 1241 | 63 | ||
XM_021838899.1 | XP_021694591.1 | 4 - 5 | 1516 | 63 | ||
XM_021838900.1 | XP_021694592.1 | 4 - 5 | 1416 | 63 | ||
XM_021838901.1 | XP_021694593.1 | 4 - 5 | 1211 | 63 | ||
XM_021838902.1 | XP_021694594.1 | 4 - 5 | 1117 | 63 | ||
XM_021838903.1 | XP_021694595.1 | 4 - 5 | 996 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
