Assay Details
Gene Symbol: LOC5574326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661307.2 | XP_001661357.2 | 6 - 7 | 1203 | 95 | |
XM_021838898.1 | XP_021694590.1 | 6 - 7 | 1502 | 95 | ||
XM_021838899.1 | XP_021694591.1 | 6 - 7 | 1777 | 95 | ||
XM_021838900.1 | XP_021694592.1 | 6 - 7 | 1677 | 95 | ||
XM_021838901.1 | XP_021694593.1 | 6 - 7 | 1472 | 95 | ||
XM_021838902.1 | XP_021694594.1 | 6 - 7 | 1378 | 95 | ||
XM_021838903.1 | XP_021694595.1 | 6 - 7 | 1257 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
