Assay Details
Gene Symbol: LOC5577718 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842485.1 | XP_021698177.1 | 3 - 4 | 2213 | 87 | |
XM_021842486.1 | XP_021698178.1 | 3 - 4 | 1741 | 87 | ||
XM_021842487.1 | XP_021698179.1 | 3 - 4 | 1671 | 87 | ||
XM_021842488.1 | XP_021698180.1 | 4 - 5 | 1913 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021842485.1, XM_021842486.1, XM_021842487.1, XM_021842488.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
