Assay Details
Gene Symbol: LOC5577718 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842485.1 | XP_021698177.1 | 1 - 2 | 564 | 71 | |
XM_021842488.1 | XP_021698180.1 | 1 - 2 | 564 | 71 | ||
XM_021842489.1 | XP_021698181.1 | 1 - 2 | 564 | 71 | ||
XM_021842490.1 | XP_021698182.1 | 1 - 2 | 564 | 71 | ||
XM_021842491.1 | XP_021698183.1 | 1 - 2 | 564 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021842485.1, XM_021842488.1, XM_021842489.1, XM_021842490.1, XM_021842491.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
