Assay Details
Gene Symbol: LOC110679692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855265.1 | XP_021710957.1 | 5 - 6 | 1955 | 64 | |
XM_021855266.1 | XP_021710958.1 | 5 - 6 | 1917 | 64 | ||
XM_021855267.1 | XP_021710959.1 | 5 - 6 | 1830 | 64 | ||
XM_021855268.1 | XP_021710960.1 | 5 - 6 | 1785 | 64 | ||
XM_021855269.1 | XP_021710961.1 | 5 - 6 | 1426 | 64 |

More Information
Set Membership: |
![]() ![]() |
