Assay Details
Gene Symbol: LOC110679692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855265.1 | XP_021710957.1 | 4 - 5 | 1809 | 73 | |
XM_021855266.1 | XP_021710958.1 | 4 - 5 | 1771 | 73 | ||
XM_021855267.1 | XP_021710959.1 | 4 - 5 | 1684 | 73 | ||
XM_021855268.1 | XP_021710960.1 | 4 - 5 | 1639 | 73 | ||
XM_021855269.1 | XP_021710961.1 | 4 - 5 | 1280 | 73 |
