Assay Details
Gene Symbol: LOC5571886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843131.1 | XP_021698823.1 | 1 - 2 | 571 | 86 | |
XM_021843132.1 | XP_021698824.1 | 1 - 2 | 571 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
