Assay Details
Gene Symbol: LOC5580226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852541.1 | XP_021708233.1 | 13 - 14 | 2213 | 77 | |
XM_021852543.1 | XP_021708235.1 | 14 - 15 | 2231 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
