Assay Details
Gene Symbol: LOC5576370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839723.1 | XP_021695415.1 | 11 - 12 | 4097 | 74 | |
XM_021839733.1 | XP_021695425.1 | 11 - 12 | 4098 | 74 | ||
XM_021839737.1 | XP_021695429.1 | 7 - 8 | 2700 | 74 | ||
XM_021839743.1 | XP_021695435.1 | 7 - 8 | 2700 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
