Assay Details
Gene Symbol: LOC5569147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658286.2 | XP_001658336.1 | 3 - 4 | 673 | 91 | |
XM_021839908.1 | XP_021695600.1 | 3 - 4 | 678 | 91 | ||
XM_021839909.1 | XP_021695601.1 | 3 - 4 | 343 | 91 | ||
XM_021839910.1 | XP_021695602.1 | 3 - 4 | 344 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
