Assay Details
Gene Symbol: LOC5569147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658286.2 | XP_001658336.1 | 1 - 2 | 359 | 75 | |
XM_021839908.1 | XP_021695600.1 | 1 - 2 | 361 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001658286.2, XM_021839908.1 transcripts.
Set Membership: |
![]() ![]() |
