Assay Details
Gene Symbol: LOC5565095 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649399.2 | XP_001649449.1 | 2 - 3 | 1070 | 86 | |
XM_021840483.1 | XP_021696175.1 | 2 - 3 | 1097 | 86 |
