Assay Details
Gene Symbol: LOC5565095 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649399.2 | XP_001649449.1 | 4 - 5 | 1347 | 83 | |
XM_021840483.1 | XP_021696175.1 | 4 - 5 | 1374 | 83 | ||
XM_021840484.1 | XP_021696176.1 | 2 - 3 | 1469 | 83 | ||
XM_021840485.1 | XP_021696177.1 | 2 - 3 | 576 | 83 |
