Assay Details
Gene Symbol: LOC5565095 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649399.2 | XP_001649449.1 | 5 - 6 | 1914 | 58 | |
XM_021840483.1 | XP_021696175.1 | 5 - 6 | 1941 | 58 | ||
XM_021840484.1 | XP_021696176.1 | 3 - 4 | 2036 | 58 | ||
XM_021840485.1 | XP_021696177.1 | 3 - 4 | 1143 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
