Assay Details
Gene Symbol: LOC5566949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838683.1 | XP_021694375.1 | 5 - 6 | 1534 | 61 | |
XM_021838684.1 | XP_021694376.1 | 5 - 6 | 1545 | 61 | ||
XM_021838685.1 | XP_021694377.1 | 4 - 5 | 903 | 61 | ||
XM_021838686.1 | XP_021694378.1 | 4 - 5 | 1044 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
