Assay Details
Gene Symbol: LOC5568108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840951.1 | XP_021696643.1 | 2 - 3 | 623 | 59 | |
XM_021840952.1 | XP_021696644.1 | 2 - 3 | 619 | 59 | ||
XM_021840953.1 | XP_021696645.1 | 2 - 3 | 503 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
