Assay Details
Gene Symbol: LOC5568785 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658100.2 | XP_001658150.2 | 1 - 2 | 200 | 73 | |
GenBank mRNA | AY432545.1 | - | 1 - 2 | 37 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
