Assay Details
Gene Symbol: LOC5576003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850280.1 | XP_021705972.1 | 1 | 73 | ||
GenBank mRNA | AY432655.1 | - | 1 | 622 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
