Assay Details
Gene Symbol: LOC5576221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655949.2 | XP_001655999.2 | 2 - 3 | 295 | 65 | |
GenBank mRNA | AY432661.1 | - | 2 - 3 | 294 | 65 | |
DQ459303.1 | - | 1 - 2 | 148 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
