Assay Details
Gene Symbol: LOC5576221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655949.2 | XP_001655999.2 | 3 - 4 | 510 | 73 | |
GenBank mRNA | AY432661.1 | - | 3 - 4 | 509 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
