Assay Details
Gene Symbol: LOC5576221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655949.2 | XP_001655999.2 | 4 - 5 | 631 | 68 | |
GenBank mRNA | AY432661.1 | - | 4 - 5 | 630 | 68 | |
DQ459303.1 | - | 3 - 4 | 484 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
