Assay Details
Gene Symbol: LOC5573994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661030.2 | XP_001661080.1 | 2 - 3 | 431 | 66 | |
GenBank mRNA | AY432693.1 | - | 2 - 3 | 417 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
