Assay Details
Gene Symbol: LOC5572845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660553.2 | XP_001660603.1 | 2 - 3 | 957 | 104 | |
XM_021850020.1 | XP_021705712.1 | 4 - 5 | 1236 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
